missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) acetylacetonate --force to the web--, 35% Pd

Product Code. 11344112 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11344112 5 g Glass bottle 5g
10042570 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344112 Supplier Thermo Scientific Acros Supplier No. 197580050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14024-61-4 | C10H14O4Pd | 304.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-61-4
Molecular Formula C10H14O4Pd
Molecular Weight (g/mol) 304.64
MDL Number MFCD00000025 MFCD00000025
InChI Key JKDRQYIYVJVOPF-FDGPNNRMSA-L
Synonym palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii
PubChem CID 53384484
IUPAC Name (E)-4-oxopent-2-en-2-olate;palladium(2+)
SMILES [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 190.0°C
Color Brown-Red
Physical Form Solution
Quantity 5 g
Linear Formula (CH3COCH=C(CH3)O)2Pd
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene
Formula Weight 304.61
Percent Purity 35% (Pd)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Palladium(II) acetylacetonate, 35% Pd
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Palladium(II) acetylacetonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.
  • P420-Store away from other materials.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.