missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) bromide, Premion™, 99.99% (metals basis), Pd 39.5% min

Product Code. p-4407684
Click to view available options
Quantity:
2 g
10 g
missing translation for 'unitSize'
10g
2g
This item is not returnable. View return policy

Product Code. 11381958

missing translation for 'mfr': Thermo Scientific Alfa Aesar 011878.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13444-94-5 | H2O2Pd | 140.43 g/mol

Palladium(II) bromide is used as an oxidizing agent, as Pd(II) catalyst and Pd(0) precatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-94-5
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
IUPAC Name palladium(2+) dihydroxide
SMILES [OH-].[OH-].[Pd++]

Specifications

Melting Point (decomposition)
Color Black
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.99% (metal basis)
Solubility Information soluble in hydrogen bromide. Insoluble in water.
Formula Weight 266.22
Odor Odorless
Sensitivity Hygroscopic
Appearance Black moist powder
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Palladium hydroxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.