missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pam3CSK4, TLR1 and TLR2 Ligand

Product Code. 18705143 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18705143 0.025 mg 25µg
18796653 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18705143 Supplier Novus Biologicals™ Supplier No. NBP2252970.025mg
  • ONLINE EXCLUSIVE PRICE
    259.00 €
    244.65 € / 25µg
    Save 14.35 €
    5% Off
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Applications: Functional, In vitro assay, In vivo assay, Ligand Activation

Pam3CSK4, TLR1 and TLR2 Ligand is a synthetic tripalmitoylated lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein. It activates monocytes and macrophages and is also a potent activator of proinflammatory transcription factor NF-kB. Pam3CSK4, TLR1 and TLR2 Ligand is recognized by a heterodimer formed between TLR1 and TLR2. Pam3CSK4, TLR1 and TLR2 Ligand stimulation: If your cell line does not naturally express TLR1+ TLR2, co-transfect with plasmids pCMV/TLR1and pCMV/TLR2. Forty-eight hours after transfection, stimulate cells with 10 to 100ng/ml of Pam3CSK4 for 6 to 24 hours. Determine Pam3CSK4 stimulation using appropriate detection assays. Buffer: Sterile water
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.025 mg
For Use With (Application) Activation of TLR1 and TLR2 heterodimer. Stimulation of TLR1 and TLR2 has been reported with 10 to 100ng/mL. Use in vitro assay reported in scientific literature (PMID: 24105263) Use in functional reported in scientific literature (PMID 25511699). Use in In-vivo reported in scientific literature (PMID 25466255). Use in ligand activation reported in multiple pieces scientific literature.
Target Pam3CSK4, TLR1 and TLR2 Ligand
Species Human/Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.