missing translation for 'onlineSavingsMsg'
Learn More

Pamidronic acid, MedChemExpress

Product Code. 16434207
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434207 Supplier Medchem Express Supplier No. HYB0012
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pamidronic acid is a drug used to treat a broad spectrum of bone absorption diseases.

Pamidronic acid is a drug used to treat a broad spectrum of bone absorption diseases.

Specifications

Chemical Name or Material Pamidronic acid
Physical Form Powder
CAS 40391-99-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C3H11NO7P2
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 5 mg/mL (21.27 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OC(P(O)(O)=O)(P(O)(O)=O)CCN
Molecular Weight (g/mol) 235.07
Formula Weight 235.07
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.