missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pan-cadherin Recombinant Rabbit Monoclonal Antibody (23GB5050), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552774
This item is not returnable.
View return policy
Description
Pan Cadherin including CDH1, CDH2, CDH3, CDH4 protein belong to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin.This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry. At certain central nervous system synapses, presynaptic to postsynaptic adhesion is mediated at least in part by this gene product.
Specifications
| Pan-cadherin | |
| Recombinant Monoclonal | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09803, P10287, P12830, P15116, P19022, P22223, P39038, P55283 | |
| CDH1, CDH2, CDH3, CDH4 | |
| A synthesized peptide derived from human pan Cadherin (850-906AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5050 | |
| Unconjugated | |
| CDH1 | |
| 2600010P09Rik; 4930525N21Rik; AA960649; AF020312; AI385538; AI851787; Arc-1; ATP-dependent helicase CHD1; ATP-dependent helicase CHD3; AW120700; AW555109; B-cadherin; CAD4; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, type 1; cadherin 1, type 1 S homeolog; cadherin 1, type 1, E-cadherin (epithelial); cadherin 15, M-cadherin (myotubule); cadherin 2; cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal); cadherin 3; cadherin 3, P-cadherin (placental); cadherin 3, type 1, P-cadherin (placental); cadherin 4; cadherin 4 type 1 R-cadherin (retinal); cadherin 4, type 1, preproprotein; cadherin 4, type 1, R-cadherin (retinal); cadherin e; Cadherin-1; Cadherin-2; cadherin-3; Cadherin-4; cadherin-E; Cadherin-N; Cadp; calcium-dependent adhesion protein, epithelial; calcium-dependent adhesion protein, neuronal; calcium-dependent adhesion protein, placental; CAM 120/80; CD324; CD325; cdh1; cdh1.S; Cdh2; Cdh3; Cdh4; Cdh4l; CDHE; CDHN; Cdhp; CDw325; CDw325 antigen; cell-CAM 120/80; CHD1; CHD-1; CHD3; CHD-3; Chd7; chromodomain helicase DNA binding protein 1; chromodomain helicase DNA binding protein 3; chromodomain-helicase-DNA-binding protein 1; Chromodomain-helicase-DNA-binding protein 3; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; HJMD; hZFH; K-CAM; LCAM; L-CAM; Liver cell adhesion molecule; liver cell adhesion protein; liver cell adhesion protein precursor; LOC108702627; Maternally-expressed neural cadherin; Mi-2 alpha; mi-2 autoantigen 240 kDa protein; Mi-2a; Mi2-ALPHA; Ncad; N-cadherin; N-cadherin 1; Neural cadherin; neural calcium-dependent cell adhesion molecule; Neural-cadherin; Pan Cadherin; PCAD; P-cadherin; P-cadherin (AA 232-723); Placental cadherin; Placental-cadherin; Prp7; Prp9-1; RCAD; R-CAD; R-Cadh; R-cadherin; retina cadherin; retinal cadherin; RPE-specific cadherin; Um; Unknown (protein for IMAGE:8237502); unnamed protein product; UVO; Uvomorulin; XELAEV_18024918mg; XELAEV_18046239mg; XmN-cadherin; xTCAD-1; ZFH; zinc finger helicase; zinc-finger helicase (Snf2-like) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1000, 1001, 1002, 12550, 12558, 12560, 12561, 999 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction