missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan-Endothelium Marker, p Ser123, p Ser124 Antibody (HIS52) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00076
This item is not returnable.
View return policy
Description
Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).
Specifications
| Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1-10 ug/ml, Western Blot (Negative) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Rat | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| HIS52 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| The immunogen for this antibody is Pan endothelium. | |
| 0.1 mg | |
| Primary | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction