missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tryptophan, 99%

Product Code. 15456199
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456199 100 g 100g
15436199 5 g 5g
15446199 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15456199 Supplier Thermo Scientific Alfa Aesar Supplier No. J64096.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1218-34-4 | C13H14N2O3 | 246.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218-34-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00065976
InChI Key DZTHIGRZJZPRDV-LBPRGKRZSA-N
Synonym n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid
PubChem CID 700653
ChEBI CHEBI:74640
IUPAC Name (2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 186°C
Color White
Quantity 100 g
Formula Weight 246.3
Optical Rotation +26° (c=1 in 1N NaOH)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-tryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.