missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole sodium, MedChemExpress

Product Code. 16457346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457346 100 mg 100mg
16411876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16457346 Supplier Medchem Express Supplier No. HY17507A100mg
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pantoprazole sodium (BY10232 sodium) is an orally active and potent proton pump inhibitor (PPI). Pantoprazole sodium, a substituted benzimidazole, is a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. Pantoprazole sodium improves pH stability and has anti-secretory, anti-ulcer activities. Pantoprazole sodium significantly increased tumor growth delay combined with Doxorubicin (HY-15142).

Pantoprazole sodium (BY10232 sodium) is an orally active and potent proton pump inhibitor (PPI). Pantoprazole sodium, a substituted benzimidazole, is a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. Pantoprazole sodium improves pH stability and has anti-secretory, anti-ulcer activities. Pantoprazole sodium significantly increased tumor growth delay combined with Doxorubicin (HY-15142).

Specifications

Chemical Name or Material Pantoprazole sodium
CAS 138786-67-1
Color White
Purity Grade Notes Research
Molecular Formula C16H14F2N3NaO4S
Quantity 100 mg
Synonym BY1023 sodiumSKF96022 sodium
Solubility Information DMSO : ≥ 100 mg/mL (246.70 mM) ∣H2O : 3.85 mg/mL (9.50 mM; Need ultrasonic)
SMILES O=S(C1=NC2=CC=C(OC(F)F)C=C2[N-]1)CC3=NC=CC(OC)=C3OC.[Na+]
Molecular Weight (g/mol) 405.35
Formula Weight 405.35
Percent Purity 99.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.