missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papain Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-44229
This item is not returnable.
View return policy
Description
Specifically detects Papain in Plant samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
Papain Polyclonal specifically detects Papain in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Papain | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Allergen Car p 1, Papaya proteinase I, PPI | |
| Papain Antibody was produced by repeated immunizations with pork Papain protein isolated from papaya. (Uniprot: P00784) | |
| Multi-step | |
| RUO | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000, Immunohistochemistry 1:500-1:3000 | |
| P00784 | |
| Goat | |
| 38.92 kDa | |
| 0.1 mg | |
| Primary | |
| Plant | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction