missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPD4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93923-0.1ml
This item is not returnable.
View return policy
Description
PAPD4 Polyclonal antibody specifically detects PAPD4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PAPD4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 2.7.7.19, FLJ38499, GLD2TUTase 2, hGLD-2, PAP associated domain containing 4, PAP-associated domain-containing protein 4, poly(A) RNA polymerase GLD2, Terminal uridylyltransferase 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PAPD4 (NP_776158.2). MFPNSILGRPPFTPNHQQHNNFFTLSPTVYSHQQLIDAQFNFQNADLSRAVSLQQLTYGNVSPIQTSASPLFRGRKRLSDEKNLPLDGKRQRFHSPHQEPTVVNQIVPLSGERRYSMPPLFHTHYVPDIVRCVPPFREIAFLEPREITLPEAKDKLSQQILELFETCQQQISDLKKKELCRTQLQREIQLLFPQSRLFLVGSSLNGFGTRSSDGDLCLVVKEEPCFFQVNQKTEARHILTLVHKHFCTRLSGYIERPQLIRAKVPIVKFRDKVSCVEFDLNVNNIVGIRNTFLLRTYAYL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 167153 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction