missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papilin Antibody (5F2D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
208.00 € - 533.00 €
Specifications
| Antigen | Papilin |
|---|---|
| Clone | 5F2D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639205
|
Novus Biologicals
NBP2-37321-0.025ml |
0.025 mL |
208.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132908
|
Novus Biologicals
NBP2-37321 |
0.1 mL |
533.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Papilin Monoclonal specifically detects Papilin in Human samples. It is validated for Western Blot, ELISA.Specifications
| Papilin | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 89932 | |
| Purified recombinant fragment of human PAPLN (AA: 766-870) expressed in E. Coli. | |
| Primary | |
| 137.7 kDa |
| 5F2D10 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Human | |
| papilin, Papilin, Proteoglycan-Like Sulfated Glycoprotein, PAPLN | |
| PAPLN | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title