Learn More
Invitrogen™ PAPPA Monoclonal Antibody (PAPP A 1-1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0060102
Description
ABS 006-01-02 detects native Pregnancy-associated plasma protein A (PAPP-A) from human samples. ABS 006-01-02 has been successfully used in Western blotting, Immunohistochemistry and ELISA procedures. By Western blot, only unreduced forms of PAPA-A can be detected. In sandwich ELISA with a polyclonal antibody in coat, ABS 006-01-02 only reacts poorly with PAPP-A in third trimester serum. The epitope of ABS 006-01-02 is different from BTE 004-09-02 and ABS 006-24-02. The epitope is located on the PAPP-A subunit of the heterotetramer. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Pregnancy-associated plasma protein-A (PAPP-A, Insulin-like growth factor binding protein-4 protease, PAPP-A-proMBP complex) is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down's syndrome (trisomy 21).
Specifications
| PAPPA | |
| Monoclonal | |
| 1.10 mg/mL | |
| PBS with 15 mM sodium azide; pH 7.4 | |
| Q13219 | |
| PAPPA | |
| Purified human PAPP-A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| PAPP A 1-1 | |
| Unconjugated | |
| PAPPA | |
| 8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5069 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.