missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (PAPP A 1-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21614-0.025ML
This item is not returnable.
View return policy
Description
Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Pappalysin-1/PAPP-A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Specific for native human PAPP-A. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| PAPP A 1-1 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide | |
| PAPPA | |
| Purified human PAPP-A adsorbed onto aluminum hydroxide gel. | |
| 0.025 mL | |
| Cancer | |
| 5069 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction