missing translation for 'onlineSavingsMsg'
Learn More

Paquinimod, MedChemExpress

Product Code. 16483018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483018 10 mg 10mg
16464326 100 mg 100mg
16459586 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483018 Supplier Medchem Express Supplier No. HY100442
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Paquinimod (ABR 25757) is a specific inhibitor of S100A8/S100A9. Paquinimod rescues the pneumonia with substantial reduction of viral loads in SARS-CoV-2-infected mice.

Paquinimod (ABR 25757) is a specific inhibitor of S100A8/S100A9. Paquinimod rescues the pneumonia with substantial reduction of viral loads in SARS-CoV-2-infected mice.

Specifications

Chemical Name or Material Paquinimod
Physical Form Solid
CAS 248282-01-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H22N2O3
For Use With (Application) COVID-19-immunoregulation
Synonym ABR 25757
Solubility Information DMSO : ≥ 125 mg/mL (356.72 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C1=C(O)C2=C(C=CC=C2CC)N(C)C1=O)N(CC)C3=CC=CC=C3
Molecular Weight (g/mol) 350.41
Formula Weight 350.41
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.