missing translation for 'onlineSavingsMsg'
Learn More

Paraffin, pure, wax, granular

Product Code. 15254734 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
Unit Size:
10kg
2kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15254734 10 kg 10kg
10711452 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15254734 Supplier Thermo Scientific Acros Supplier No. 416770100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure | CAS: 8002-74-2 | CnH2n+2 | 341.451 g/mol

Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-74-2
Molecular Formula CnH2n+2
Molecular Weight (g/mol) 341.451
MDL Number MFCD00132833
InChI Key JWHAUXFOSRPERK-UHFFFAOYSA-N
Synonym propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn
PubChem CID 4932
ChEBI CHEBI:63619
IUPAC Name 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
SMILES CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O

Specifications

Melting Point 58.0°C to 62.0°C
Color White
Boiling Point 322.0°C
Flash Point 198°C
Infrared Spectrum Authentic
Quantity 10 kg
Merck Index 14,7023
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alcohol (soluble in warm alcohol),insoluble in acids,soluble in benzene,chloroform,ether,carbon,disulfide,ligroin,turpentine,miscible when melted with wax,fats
Viscosity 3-4.5 mPa.s (98.9°C)
Grade Pure
Physical Form Pellets
Chemical Name or Material Paraffin wax
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.