missing translation for 'onlineSavingsMsg'
Learn More

Paraffin Wax, Pure, Melting Point about 60°C, Pastillated, Fisher Chemical

Product Code. 10395900 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
450 g
Packaging:
Plastic powder jar
Unit Size:
450g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10395900 450 g Plastic powder jar 450g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10395900 Supplier Fisher Chemical Supplier No. P/0680/90
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

450g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 8002-74-2
Melting Point 58°C
Color White
Boiling Point 322°C
Packaging Plastic powder jar
Molecular Formula C21H27NO3
MDL Number 132833
Quantity 450 g
Synonym propafenone, rythmol, propafenona, propafenonum, propafenonum inn-latin, propafenona inn-spanish, propafenone inn:ban, gnf-pf-4594, propafenone-hcl, propafenone inn
InChI Key JWHAUXFOSRPERK-UHFFFAOYSA-N
SMILES CCCNCC(COC1=CC=CC=C1C(=O)CCC2=CC=CC=C2)O
IUPAC Name 1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylpropan-1-one
Molecular Weight (g/mol) 341.451
PubChem CID 4932
ChEBI CHEBI:63619
Viscosity 3 to 4.5 mPaS at 98.9°C
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.