missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline Base, TRC

Product Code. 30136708
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136708 10 g 10g
17982165 5 g 5g
17914755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136708 Supplier Toronto Research Chemicals Supplier No. TRCP19328810G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 467-62-9
Molecular Formula C19 H19 N3 O
Molecular Weight (g/mol) 305.37
Synonym Parafuchsin Carbinol, 4-Amino-α,α-bis(4-aminophenyl)benzenemethanol, Tris(4-aminophenyl)methanol, Tris(p-aminophenyl)methanol
IUPAC Name tris(4-aminophenyl)methanol
SMILES Nc1ccc(cc1)C(O)(c2ccc(N)cc2)c3ccc(N)cc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H19N3O/c20-16-7-1-13(2-8-16)19(23,14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15/h1-12,23H,20-22H2
Formula Weight 305.1528
Chemical Name or Material Pararosaniline Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.