missing translation for 'onlineSavingsMsg'
Learn More

Parathion, TRC

Product Code. 30133339
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133339 500 mg 500mg
30040996 100 mg 100mg
30041389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133339 Supplier Toronto Research Chemicals Supplier No. TRCP192220500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-38-2
Molecular Formula C10 H14 N O5 P S
Molecular Weight (g/mol) 291.26
Synonym Phosphorothioic acid, O,O-diethyl O-(4-nitrophenyl) ester, Phosphorothioic acid, O,O-diethyl O-(p-nitrophenyl) ester (6CI,8CI), AAT, AATP, Alkron, Alleron, American Cyanamid 3422, Aphamite, Aralo, BAY 1605, Bayer E-605, Bladan F, DNTP, Diethyl 4-nitrophenyl phosphorothioate, Diethyl p-nitrophenyl phosphorothionate, Diethyl p-nitrophenyl thionophosphate, Diethyl parathion, E 605, E 605F, E 605FT20, ENT 15108, Ecatox 20, Ekatox, Ekatox 20, Ethyl parathion, Etilon, Folidol, Folidol E, Folidol E-605, Folidol oil, Fosferno, Fostox, Galpar, Gearphos, Lirothion, NIUIF 100, NSC 8933, Niran, Nitrostigmine, Nourithion, O,O-Diethyl O-(4-nitrophenyl) phosphorothioate, O,O-Diethyl O-(p-nitrophenyl) phosphorothioate, O,O-Diethyl O-p-nitrophenyl thiophosphate, Oleofos 20, Oleoparathene, Oleoparathion, Pacol, Paramar 50, Paraphos, Parathene, Parathion, Parathion A, Parathion-ethyl, Penncap E, RB, Rhodiasol, Rhodiatox, Selephos, Super Rodiatox, Thiomex, Thiophos, Thiophos 3422, Vitrex
IUPAC Name diethoxy-(4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1ccc(cc1)[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H14NO5PS/c1-3-14-17(18,15-4-2)16-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3
Formula Weight 291.033
Percent Purity >95%
Chemical Name or Material Parathion-ethyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.