missing translation for 'onlineSavingsMsg'
Learn More

Parbendazole, MedChemExpress

Product Code. 16485636
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485636 100 mg 100mg
16471107 50 mg 50mg
16404858 10 mg 10mg
16403266 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485636 Supplier Medchem Express Supplier No. HY115364
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Parbendazole is a potent inhibitor of microtubule assembly, destabilizes tubulin, with an EC50 of 530 nM, and exhibits a broad-spectrum anthelmintic activity.

Parbendazole is a potent inhibitor of microtubule assembly, destabilizes tubulin, with an EC50 of 530 nM, and exhibits a broad-spectrum anthelmintic activity.

Specifications

Chemical Name or Material Parbendazole
Physical Form Solid
CAS 14255-87-9
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H17N3O2
For Use With (Application) COVID-19-anti-virus
Synonym SKF 29044
Solubility Information DMSO : 4 mg/mL (16.18 mM; Need ultrasonic)
SMILES O=C(OC)NC1=NC2=CC=C(CCCC)C=C2N1
Molecular Weight (g/mol) 247.29
Formula Weight 247.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.