missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pardoprunox hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Pardoprunox (SLV-308) hydrochloride is a partial dopamine D2 and D3 receptor partial agonist and a serotonin 5-HT1A receptor agonist, with pEC50s of 8, 9.2, and 6.3, respectively.
Specifications
Specifications
| Chemical Name or Material | Pardoprunox hydrochloride |
| CAS | 269718-83-4 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C12H16ClN3O2 |
| Quantity | 50 mg |
| Synonym | SLV-308 hydrochloride DU-126891 hydrochloride |
| Solubility Information | DMSO : 150 mg/mL (556.11 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1OC2=C(N3CCN(C)CC3)C=CC=C2N1.[H]Cl |
| Molecular Weight (g/mol) | 269.73 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction