missing translation for 'onlineSavingsMsg'
Learn More

Parecoxib, MedChemExpress

Product Code. 16457326
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457326 100 mg 100mg
16443576 25 mg 25mg
16433387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16457326 Supplier Medchem Express Supplier No. HY17474
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Parecoxib (SC 69124) is a highly selective and orally active COX-2 inhibitor, the prodrug of Valdecoxib (HY-15762). Parecoxib Sodium is a nonsteroidal anti-inflammatory agent (NSAID) and inhibits prostaglandin (PG) synthesis. Parecoxib can be used for the relief of acute postoperative pain and symptoms of chronic inflammatory conditions such as osteoarthritis and rheumatoid arthritis in vivo.

Parecoxib (SC 69124) is a highly selective and orally active COX-2 inhibitor, the prodrug of Valdecoxib (HY-15762). Parecoxib Sodium is a nonsteroidal anti-inflammatory agent (NSAID) and inhibits prostaglandin (PG) synthesis. Parecoxib can be used for the relief of acute postoperative pain and symptoms of chronic inflammatory conditions such as osteoarthritis and rheumatoid arthritis in vivo.

Specifications

Chemical Name or Material Parecoxib
CAS 198470-84-7
Color White
Purity Grade Notes Research
Molecular Formula C19H18N2O4S
Quantity 100 mg
Synonym SC 69124
Solubility Information DMSO : ≥ 50 mg/mL (134.98 mM)
SMILES CCC(NS(=O)(C1=CC=C(C2=C(C)ON=C2C3=CC=CC=C3)C=C1)=O)=O
Molecular Weight (g/mol) 370.42
Formula Weight 370.42
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.