missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARK2/Parkin Mouse anti-Human, Mouse, Clone: 2H5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66674-1-IG-20UL
This item is not returnable.
View return policy
Description
The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.Specifications
| PARK2/Parkin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60260 | |
| Park2, PRKN | |
| PARK2/Parkin Fusion Protein Ag5179 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 2H5A7 | |
| Unconjugated | |
| PRKN | |
| AR JP, LPRS2, PARK2, Parkinson disease protein 2, PDJ, PRKN | |
| Mouse | |
| Protein A | |
| RUO | |
| 5071, 50873 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction