missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83592-25ul
This item is not returnable.
View return policy
Description
PARP12 Polyclonal specifically detects PARP12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PARP12 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9H0J9 | |
| PARP12 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AATLKFQAGKHNYELDFKAFVQKNLVYGTTKKVCRRPKYVSPQDVTTMQTCNTKFPGPKSIPDYWDSSALPDPGFQKITLSSSSEEYQKVWNLFNRTLPFYFVQKIERVQNLALWEVYQWQKGQMQKQNGG | |
| 25 μL | |
| Apoptosis, DNA Repair, Hypoxia | |
| 64761 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 2.4.2.30, FLJ22693MST109, PARP-12, poly (ADP-ribose) polymerase family, member 12, poly [ADP-ribose] polymerase 12, ZC3H1, ZC3HDC1MSTP109, Zinc finger CCCH domain-containing protein 1, zinc finger CCCH type domain containing 1, zinc finger CCCH-type domain containing 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction