missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
198.00 € - 522.00 €
Specifications
| Antigen | Parvalbumin |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607937
|
Novus Biologicals
NBP2-50036 |
0.1 mL |
522.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640818
|
Novus Biologicals
NBP2-50036SS |
0.025 mL |
198.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Parvalbumin Polyclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry Free-FloatingSpecifications
| Parvalbumin | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
| Unconjugated | |
| Chicken | |
| Neuroscience | |
| Supplied as an aliquot of IgY preparation | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Recombinant full length human Parvalbumin. [UniProt# P20472] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P20472, P20472 | |
| 5816 | |
| IgY | |
| IgY purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title