missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvovirus VP2 Antibody (4E5A4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61902
This item is not returnable.
View return policy
Description
Parvovirus VP2 Monoclonal specifically detects Parvovirus VP2 in Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Parvovirus VP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Parvovirus capsid protein 2, Parvovirus major capsid protein | |
| Mouse | |
| 46.5 kDa | |
| 0.1 mL | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4E5A4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| VP2 | |
| Purified recombinant fragment of human VP2 (AA: 296-438) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction