missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PATE4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15537212
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15537212 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15537212 Supplier Invitrogen™ Supplier No. PA546582

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: KENELCSTTA YFRGDKHMYS THMCKYKCRE EESSKRGLLR VTLCCDRNFC Sequence homology: Human: 100%.

PATE4 may modulate the function of nicotinic acetylcholine receptors and may enhance sperm motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PATE4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PATE4
Gene Accession No. P0C8F1
Gene Alias PATE4; PATE-B; PATE-like protein B; prostate and testis expressed 4; prostate and testis expressed protein 4; prostate and testis expression B
Gene Symbols PATE4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PATE4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399968
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.