Learn More
Pax2 Antibody (CL9469) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88934AF700
Product Code. 30304519
  • 737.00 € / 0.10mL
Description
Pax2 Monoclonal antibody specifically detects Pax2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pax2
Monoclonal
Alexa Fluor 700
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2
This antibody was developed using a synthetic peptide derived from Q02962, with the exact immunogen sequence remaining proprietary.
0.1 mL
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers
5076
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL9469
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified