missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAX6 Monoclonal Antibody (13B10-1A10)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA1109

361.50 EUR valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.


Product Code. 15547191

  • 482.00 € / 100µg
Seek Out Great Savings With the Lab of Curiosity
Use promo code 24110
View all promotions

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

Western blot analysis of MA1-109 detects an ~47 kDa protein in whole cell lysates from early passage human B3 lens epithelial cells and in mouse eye lysate. An additional unknown band at ~36 kDa is also detected in B3 cell lysates. This antibody will detect additional nonspecific bands in lysates with very low basal PAX6 expression. Specificity of the antibody was confirmed in B3 and mouse eye lysates (positive controls) compared to U-2 OS human osteosarcoma and NIH/3T3 mouse embryonic fibroblasts (negative controls). MA1-109- can be used for immunofluoresence and flow cytometry analysis of Pax6 in human neural stem cells.

This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter's anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAX6
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
P26367, P63015
PAX6
Purified internal fragment of human recombinant PAX6 expressed in E. coli.
100 μg
Primary
Human, Mouse
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
13B10-1A10
Unconjugated
PAX6
1500038E17Rik; AEY11; AN; AN2; Aniridia; aniridia type II protein; D11S812E; Dey; Dickie's small eye; FVH1; Gsfaey11; Keratitis; MGC17209; MGDA; oculorhombin; paired box 6; paired box gene 6; Paired Box Gene-6; paired box homeotic gene 6; paired box homeotic gene-6; paired box protein Pax-6; Pax6; Pax-6; PAX6 protein; Sey; small eye; transcription factor; WAGR
Mouse
Protein A
RUO
18508, 5080
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.