missing translation for 'onlineSavingsMsg'
Learn More

PBIT, MedChemExpress

Product Code. 16456097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456097 5 mg 5mg
16403378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16456097 Supplier Medchem Express Supplier No. HY101451
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PBIT is a specific inhibitor of the Jumonji AT-rich Interactive Domain 1 (JARID1) enzymes. PBIT inhibits JARID1B (KDM5B or PLU1) histone demethylase with an IC50 of about 3 μM . PBIT also inhibits JARID1A and JARID1C with IC50s of 6 μM and 4.9 μM, respectively.

PBIT is a specific inhibitor of the Jumonji AT-rich Interactive Domain 1 (JARID1) enzymes. PBIT inhibits JARID1B (KDM5B or PLU1) histone demethylase with an IC50 of about 3 μM . PBIT also inhibits JARID1A and JARID1C with IC50s of 6 μM and 4.9 μM, respectively.

Specifications

Chemical Name or Material PBIT
Physical Form Solid
CAS 2514-30-9
Color Brown
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C14H11NOS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (207.20 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=C(C)C=C2)SC3=C1C=CC=C3
Molecular Weight (g/mol) 241.31
Formula Weight 241.31
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.