missing translation for 'onlineSavingsMsg'
Learn More

PBS 10X, MP Biomedicals™

Product Code. 11407130 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407130 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11407130 Supplier MP Biomedicals Supplier No. 11PBS10X02

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

PBS 10X Rnase, Dnase, Protease free, Filtered through 0,2μm PTFE.

  • PBS 10 X buffer is ideal for maintaining a constant pH, and since it is isotonic and non-toxic to cells, it can be used in a wide variety of biological applications.
  • PBS 10X is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.06 g/mL
For Use With (Application) Suitable for cell culture
pH 7.4
Physical Form Liquid
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.