missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS (10X), pH 7.2
GREENER_CHOICE

Product Code. p-7076136
Click to view available options
Quantity:
500mL
10 x 500 mL
Unit Size:
5000mL
500mL
This item is not returnable. View return policy

Product Code. 11514526

Brand: Gibco™ 70013016

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

PBS (10X), pH 7.2

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ PBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ PBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
For Use With (Application) Mammalian Cell Culture
pH 7.1 to 7.3
Physical Form Liquid
Quantity 500mL
Osmolality 2800 to 3200 mOsm/kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.