missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents
34.70 € - 127.00 €
Specifications
Color | White |
---|---|
Content And Storage | Room temperature |
Packaging Type | Poly Bottle |
Form | Powder |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
10316743
|
Fisher Bioreagents
BP661-10 |
98.9 grams (makes 10L solution) | Poly Bottle |
34.70 €
98.90g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10562765
|
Fisher Bioreagents
BP661-50 |
492 grams (makes 50L solution) | Poly Bottle |
127.00 €
492g |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This product provides a convenient way to make large volumes of standard 1X Phosphate Buffer; each bottle contains 98.9g or 492g for reconstitution in 10L or 50L of water, respectively.Specifications
White | |
Room temperature | |
Poly Bottle | |
Powder |