missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCBP4 Recombinant Mouse Monoclonal Antibody (K1E075_14F9)

Product Code. 30830056
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830056 100 μL
30830390 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830056 Supplier Invitrogen™ Supplier No. MA564494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: EPELSITLTL RMLMHGKEVG SIIGKKGETV KRIREQSSAR ITISEGSCPE RITTITGSTA AVFHAVSMIA FKLDEDLCAA PANGGNVSRP PVTLRLVIPA SQCGSLIGKA GTKIKEIRET TGAQVQVAGD LLPNSTERAV TVSGVPDAII LCVRQICAVI LESPPKGATI PYHPSLSLGT VLLSANQGFS VQGQYGAVTP AEVTKLQQLS SHAVPFATPS VVPGLDPGTQ TSSQEFLVPN DLIGCVIGRQ GSKISEIRQM SGAHIKIGNQ AEGAGERHVT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 32%, Pig - 99%, Cynomolgus monkey - 100%.

MCG10 is a member of the KH-domain RNA-binding protein family. Proteins of this family also referred to as alpha-CPs, bind to RNA with specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. MCG10 is induced by p53 and DNA damage via two p53-responsive promoter elements. Its overexpression in transfected cells results in the suppression of cell proliferation by inducing apoptosis and cell cycle arrest in G2-M. Decreased expression of one isoform of MCG10 has been observed in many forms of lung cancer, leading to speculation that MCG10 can act as a tumor suppressor. Multiple alternatively spliced isoforms of MCG10 have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCBP4
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone K1E075_14F9
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene PCBP4
Gene Accession No. P57723
Gene Alias 1200003L19Rik; alpha CP-4; alpha-CP4; AlphaCP-4; alphaCP-4 protein; CBP; LIP4; LYST-interacting protein; MCG10; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4; RNA binding protein MCG10
Gene Symbols PCBP4
Host Species Mouse
Immunogen Recombinant human PCBP4 (aa 11-290).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57060
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.