Learn More
Invitrogen™ PCBP4 Recombinant Mouse Monoclonal Antibody (K1E075_14F9)
Description
Immunogen sequence: EPELSITLTL RMLMHGKEVG SIIGKKGETV KRIREQSSAR ITISEGSCPE RITTITGSTA AVFHAVSMIA FKLDEDLCAA PANGGNVSRP PVTLRLVIPA SQCGSLIGKA GTKIKEIRET TGAQVQVAGD LLPNSTERAV TVSGVPDAII LCVRQICAVI LESPPKGATI PYHPSLSLGT VLLSANQGFS VQGQYGAVTP AEVTKLQQLS SHAVPFATPS VVPGLDPGTQ TSSQEFLVPN DLIGCVIGRQ GSKISEIRQM SGAHIKIGNQ AEGAGERHVT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 32%, Pig - 99%, Cynomolgus monkey - 100%.
Specifications
Specifications
| Antigen | PCBP4 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K1E075_14F9 |
| Concentration | 0.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | PCBP4 |
| Gene Accession No. | P57723 |
| Gene Alias | 1200003L19Rik; alpha CP-4; alpha-CP4; AlphaCP-4; alphaCP-4 protein; CBP; LIP4; LYST-interacting protein; MCG10; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4; RNA binding protein MCG10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.