missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93165-0.1ml
This item is not returnable.
View return policy
Description
PCDHA13 Polyclonal antibody specifically detects PCDHA13 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PCDHA13 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CNR5, CNRN5, CNRS5, CRNR5, PCDH-ALPHA13, protocadherin alpha 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-360 of human PCDHA13 (NP_061727.1). GEIRTKGKLDFEEKKLYEISVEAVDKGNIPMAGHCTLLVEVLDVNDNAPEVTITSLSLPIR | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 56136 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction