missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHB15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00 € - 322.00 €
Specifications
| Antigen | PCDHB15 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654748
|
Novus Biologicals
NBP3-12574-100ul |
100 μg |
322.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623278
|
Novus Biologicals
NBP3-12574-50ul |
50 μg |
183.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCDHB15 Polyclonal antibody specifically detects PCDHB15 in Human samples. It is validated for Western BlotSpecifications
| PCDHB15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| PCDH-beta-15, PCDH-BETA15, protocadherin beta 15, protocadherin beta-15 | |
| Produced in rabbits immunized with E. coli-derived Human PCDHB15 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 56121 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title