missing translation for 'onlineSavingsMsg'
Learn More

PCI-27483, MedChemExpress

Product Code. 16479587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479587 5 mg 5mg
16427048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479587 Supplier Medchem Express Supplier No. HY16382
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PCI-27483 is a FVIIa/tissue factor inhibitor, with antitumour effects.

PCI-27483 is a FVIIa/tissue factor inhibitor, with antitumour effects.

Specifications

Chemical Name or Material PCI-27483
CAS 871266-63-6
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C26H24N6O9S
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (104.77 mM; Need ultrasonic)
SMILES O=C(O)C[C@@H](C(O)=O)NC(CC1=CC(C2=CC(S(=O)(N)=O)=CC=C2O)=C(O)C(C3=NC4=CC=C(C(N)=N)C=C4N3)=C1)=O
Molecular Weight (g/mol) 596.57
Formula Weight 596.57
Percent Purity 95.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.