missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ pCL-10A1 Retrovirus Packaging Vector

Product Code. 18736473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18736473 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18736473 Supplier R&D Systems™ Supplier No. NBP229542
  • ONLINE EXCLUSIVE PRICE
    454.26 €
    386.40 € / 10µg
    Save 67.86 €
    14% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: Retroviral Production

The pCL-10A1 packaging vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. By introducing a retroviral vector into a sell expressing retroviral proteins, retroviral particles (virions) are shed into the culture medium at the rate of at the rate of about 1 infectious particle/cell/day Retrovirus tropism is determined at 3 levels. The first is simply a function of viral envelope protein, gp70. The envelope determines which cells the virus will enter. pCL-10A1 expresses 10A1 envelope protein that was isolated from a mouse infected with amphotropic MuLV (2,3). Retroviruses obtained by cotransfection with pCL-10A1 vector will infect most mammalian cells including hamster cells.

  • Species: Human
  • Specificity: Ectotropic (usually) (MoMuLV) Mouse and rat cells only (not human), Amphotropic (from 4070A MuLV) Most mammalian cells (but not hamster) 10A1 (MuLV) Most mammalian cells (including hamster)
  • For large scale production, this plasmid may be amplified on LB plates containing 50μg/mL ampicillin
  • Buffer: 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Storage Buffer 10 μg in 1 x TE (10 mM Tris, pH 7.5, 1 mM EDTA)
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Species Mouse, Mammal
Concentration 0.3 mg/mL
Quantity 10 μg
Vector pCL-10A1 Retrovirus Packaging Vector
Product Line Novus
For Use With (Application) AMPLIFICATION: The Plasmid DNA Contains an Ampicillin Resistant Gene. For Large Scale Production, This Plasmid May be Amplified on LB Plates Containing 50 ug/ml Ampicillin. The Liquid Cultures May be Grown (at 100 ug Ampicillin/ml. Use in Retroviral Production Reported in Multiple Pieces of Scientific Literature.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.