missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ pCL-Ampho Retrovirus Packaging Vector

Product Code. 18746853 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18746853 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18746853 Supplier R&D Systems™ Supplier No. NBP229541
  • ONLINE EXCLUSIVE PRICE
    542.25 €
    442.05 € / 10µg
    Save 100.20 €
    18% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: Retroviral Production

The pCL-Ampho packaging vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. By introducing a retroviral vector into a cell expressing retroviral proteins, retroviral particles (virions) are shed into the culture medium at the rate of 1 infectious particle/cell/day. Retrovirus tropism is determined at three levels. The first is simply a function of viral envelope protein, gp70. The envelope determines which cells the virus will enter. gp70 comes in three different flavors for gene therapy. The plasmid DNA can be amplified by transforming bacteria and growing them on LB/ampicillin (100 ug/ml) media.

  • Species: Human
  • Specificity: Ectotropic (usually) (MoMuLV) Mouse and rat cells only (not human), Amphotropic (from 4070A MuLV) Most mammalian cells (but not hamster) 10A1 (MuLV) Most mammalian cells (including hamster)
  • Buffer: 10μg in 20μL sterile water

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Storage Buffer 10µg in 20µL sterile water
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 10 μg
Vector pCL-Ampho Retrovirus Packaging Vector
Product Line Novus
For Use With (Application) Use in ICC/IF Reported in Scientific Literature (PMID:34082772) For Amplification, Transform a Suitable E. Coli Host Such as HB101 or DH-5 Alpha and Plate on LB Plates Containing 50-ug/mL Ampicillin. For Plasmid DNA Preparation Start Culture From a Single Colony in LB Medium Containing 100 ug/mL Ampicillin. Use in Retroviral Production Reported in Scientific Literature (PMID: 24311986).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.