missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ pCL-Eco Retrovirus Packaging Vector

Product Code. 18736093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18736093 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18736093 Supplier R&D Systems™ Supplier No. NBP229540
  • ONLINE EXCLUSIVE PRICE
    487.00 €
    392.70 € / 10µg
    Save 94.30 €
    19% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: Retroviral Production

The pCL-ECO packaging vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. By introducing a retroviral vector into a sell expressing retroviral proteins, retroviral particles (virions) are shed into the culture medium at the rate of about 1 infectious particle/cell/day. Retrovirus tropism is determined at 3 levels. The first is simply a function of viral envelope protein, gp70. The envelope determines which cells the virus will enter. gp70 comes in three different flavors for gene therapy. Retroviruses obtained by cotransfection with pCL-Eco vector will infect mouse and rat cells, but not human cells.

  • Species: Human, Mouse, Rat
  • Specificity: Ectotropic (usually) (MoMuLV) Mouse and rat cells only (not human), Amphotropic (from 4070A MuLV) Most mammalian cells (but not hamster) 10A1 (MuLV) Most mammalian cells (including hamster)
  • For large scale production, this plasmid may be amplified on LB plates containing 50μg/mL ampicillin
  • Buffer: 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
TRUSTED_SUSTAINABILITY

Specifications

Storage Buffer 10µg in 20µL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 10 μg
Vector pCL-Eco Retrovirus Packaging Vector
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.