missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ pCLXSN Retrovirus Expression Vector

Product Code. 18110613 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18110613 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18110613 Supplier R&D Systems™ Supplier No. NBP229500
  • ONLINE EXCLUSIVE PRICE
    345.81 €
    286.65 € / 10µg
    Save 59.16 €
    17% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. pCLXSN expression vector is part of RetroMax system, designed for maximal virus titer in 293 cells

The pCLXSN expression vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. All members of the RetroMax expression vector family (pCLXSN, pCLNCX, pCLNRX, and pCLNDX) have an extended packaging signal (omega+) and are derived from safety modified retrovirus vectors in which the gag open reading frame has been stopped by a point mutation, thereby minimizing the opportunity for replication competent retrovirus production by recombination with packaging genome.

  • RetroMax system takes advantages of two properties of 293 cells (high level of transfectibility and strong E1A-mediated stimulation of CMV promoter controlled transcription)
  • 293 cells of nonmurine origin avoid problem of selective packaging and transfer of VL30 genomes (present in all murine packaging cells)
  • Vector supernatants are free of helper virus and are of sufficiently high titer within 2 days of transient transfection in 293 cells
  • For amplification, transform DH-5a or similar bacteria and plate on LB plates containing 50mg/mL ampicillin
  • Buffer: 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Storage Buffer 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 10 μg
Vector pCLXSN Retrovirus Expression Vector
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.