Learn More
PCPE-1/PCOLCE Antibody (CL6567) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76506AF532
Product Code. 30292135
  • 737.00 € / 0.10mL
Description
PCPE-1/PCOLCE Monoclonal antibody specifically detects PCPE-1/PCOLCE in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCPE-1/PCOLCE
Monoclonal
Alexa Fluor 532
PCPE, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein
This antibody was developed using a recombinant protein derived from Q15113, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
5118
Store at 4⪚C in the dark.
IgG1
Western Blot
CL6567
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified