Learn More
Thermo Scientific™ PCR Plate, 96-well, non-skirted, Sustain™ Series
Product Code. 30179682 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 plates
Unit Size:
Pack of 25
Product Code. Quantity Unit Size
30179682 25 plates Pack of 25
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 30179682 Supplier Thermo Scientific™ Supplier No. AB0600SS
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
The Thermo Scientific PCR Plate, 96-well, non-skirted, Sustain Series, is for use in PCR and qPCR applications and has a 0.3 mL well volume when paired with adhesive films or foils.
The Thermo Scientific PCR Plate, 96-well, non-skirted, Sustain Series, is for use in PCR and qPCR applications and has a 0.3 mL well volume when paired with adhesive films or foils. The plate features a cut corner located at H1 and is compatible with most thermal cyclers.
Features and benefits
  • Non-skirted format compatible with most thermal cyclers
  • Clear well bottom for sample visibility
  • Cut corner: H1
  • 0.3 mL maximum well volume (when used with adhesive or heat seals)
  • Sealable using adhesive films and foils, heat seals, and flat and domed cap strips
A breakthrough in sustainable PCR technology
Thermo Scientific Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
  • Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
  • Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
  • Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific PCR plastics
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Format SBS Standard
Autoclavable Autoclavable
Color Natural
Material Polypropylene
No. of Wells 96
Green Features ISCC PLUS
Quantity 25 plates