missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCYT1A Recombinant Rabbit Monoclonal Antibody (K01_1P86)

Product Code. 30830428
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830428 50 μL
30830119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830428 Supplier Invitrogen™ Supplier No. MA565077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PCYT1A Recombinant Monoclonal Antibody for Western Blot

This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCYT1A
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1P86
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene PCYT1A
Gene Accession No. P49585
Gene Alias CCT A; CCTA; Cctalpha; CCT-alpha; choline phosphate cytidylyltransferase 1 alpha; choline-phosphate cytidylyltransferase A; CT; CT A; CTA; CTalpha; CTP; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; Ctpct; Cttalpha; PCYT1; PCYT1A; phosphate cytidylyltransferase 1, choline, alpha; phosphate cytidylyltransferase 1, choline, alpha isoform; phosphorylcholine transferase A; SMDCRD
Gene Symbols PCYT1A
Host Species Rabbit
Immunogen A synthetic peptide of human CT A.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5130
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.