Learn More
Invitrogen™ PD-1 (CD279) Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559735
 View more versions of this product
Product Code. 30283539
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term UCB patent anti-PD-1.
Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of danger signals such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-1 (CD279) Humanized Biosimilar
Recombinant Monoclonal
Unconjugated
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG4SP