missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD 169316, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PD 169316 is a potent, cell-permeable and selective p38 MAP kinase inhibitor, with IC50 of 89 nM. PD169316 selectively inhibits the kinase activity of the phosphorylated p38 without hindering upstream kinases to phosphorylate p38. PD169316 shows antiviral activity against Enterovirus71. PD169316 shows antiviral activity against Enterovirus71.
Specifications
Specifications
| Chemical Name or Material | PD 169316 |
| Physical Form | Solid |
| CAS | 152121-53-4 |
| Color | Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H13FN4O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 12.5 mg/mL (34.69 mM; Need ultrasonic) |
| SMILES | FC(C=C1)=CC=C1C2=C(C3=CC=NC=C3)NC(C4=CC=C([N+]([O-])=O)C=C4)=N2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction