missing translation for 'onlineSavingsMsg'
Learn More

PD 169316, MedChemExpress

Product Code. p-200100806
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16444058

Brand: Medchem Express HY10578

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PD 169316 is a potent, cell-permeable and selective p38 MAP kinase inhibitor, with IC50 of 89 nM. PD169316 selectively inhibits the kinase activity of the phosphorylated p38 without hindering upstream kinases to phosphorylate p38. PD169316 shows antiviral activity against Enterovirus71. PD169316 shows antiviral activity against Enterovirus71.

PD 169316 is a potent, cell-permeable and selective p38 MAP kinase inhibitor, with IC50 of 89 nM. PD169316 selectively inhibits the kinase activity of the phosphorylated p38 without hindering upstream kinases to phosphorylate p38. PD169316 shows antiviral activity against Enterovirus71. PD169316 shows antiviral activity against Enterovirus71.

Specifications

Chemical Name or Material PD 169316
Physical Form Solid
CAS 152121-53-4
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H13FN4O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 12.5 mg/mL (34.69 mM; Need ultrasonic)
SMILES FC(C=C1)=CC=C1C2=C(C3=CC=NC=C3)NC(C4=CC=C([N+]([O-])=O)C=C4)=N2
Molecular Weight (g/mol) 360.34
Formula Weight 360.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.