missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (MIH5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-43262-0.5mg
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, In vivo assay.
Specifications
| PD-L1/B7-H1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500 | |
| anti-PD-L1, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1 blocking, PD-L1 ihc, PD-L1 immunohistochemistry, PD-L1 western blot, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a λ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| MIH5 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD274 | |
| The immunogen for this antibody was B7H1. | |
| 0.5 mg | |
| Signal Transduction | |
| 29126 | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction