missing translation for 'onlineSavingsMsg'
Learn More

PD98059, MedChemExpress

Product Code. 16484928
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484928 10 mg 10mg
16475706 100 mg 100mg
16451177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16484928 Supplier Medchem Express Supplier No. HY12028
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PD98059 is a potent and selective MEK inhibitor with an IC50 of 5 μM.

PD98059 binds to the inactive form of MEK, thereby preventing the activation of MEK1 (IC50 of 2-7 μM) and MEK2 (IC50 of 50 μM) by upstream kinases. PD98059 is a ERK1/2 signaling inhibitor. PD98059 is a ligand for the aryl hydrocarbon receptor (AHR), and suppresses TCDD binding (IC50 of 4 μM) and AHR transformation (IC50 of 1 μM). PD98059 also inhibits autophagy.

Specifications

Chemical Name or Material PD98059
CAS 167869-21-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13NO3
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (124.70 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C=C(OC2=CC=CC=C21)C3=CC=CC(OC)=C3N
Molecular Weight (g/mol) 267.28
Formula Weight 267.28
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.