missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95162-0.02ml
This item is not returnable.
View return policy
Description
PDE1C Polyclonal antibody specifically detects PDE1C in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PDE1C | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calmodulin-dependent (70kD), EC 3.1.4, phosphodiesterase 1C, calmodulin-dependent 70kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 435-634 of human PDE1C (NP_001177985.1). VEPTFTVLTDMTEKIVSPLIDETSQTGGTGQRRSSLNSISSSDAKRSGVKTSGSEGSAPINNSVISVDYKSFKATWTEVVHINRERWRAKVPKEEKAKKEAEEKARLAAEEQQKEMEAKSQAEEGASGKAEKKTSGETKNQVNGTRANKSDNPRGKNSKAEKSSGEQQQNGDFKDGKNKTDKKDHSNIGNDSKKTDDSQE | |
| 0.02 mL | |
| Signal Transduction | |
| 5137 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction